Check Order Status
Chill-Its™ 6665 Evaporative Cooling Vest
Actual product may vary
Fisherbrand
Chill-Its™ 6665 Evaporative Cooling Vest
Evaporative Cooling Vest provides cooling relief and comfort once soaked in cold water.
Have Questions?
Catalog Number
Color
Size
FIS12-547
Gray
3X-Large
Catalog number
FIS12-547
Price (MYR)
298.00
PK
Add to cart
Color:
Gray
Size:
3X-Large
Price (MYR)
298.00
PK
Add to cart
V-neck with zipper closure
Quilted nylon outer layer
Activated, polymer-embedded fabric inside
Water-repellent liner and black cotton-poly elastic trim
Lightweight, durable, and washable
Soak in water 2-5 minutes to activate
Remains hydrated for up to 4 hours
Mesh side panels offer improved ventilation and stretch
Specifications
Material
Nylon
Product Type
Vest
Size
3X-Large
Color
Gray
For Use With (Application)
Construction, Maintenance, Landscaping, Material Handling