Premium Full-Frosted Cytology Microscope Slides
Actual product may vary
Fisherbrand
Premium Full-Frosted Cytology Microscope Slides
With sandblasted frosting over one entire side
Have Questions?
Catalog number FIS12-544-5CY
Price (MYR)
-
With sandblasted frosting over one entire side
  • Frosting disappears when cover slip is mounted over specimen—doesn't interfere with viewing
  • 0.097 to 1.07mm thick
Specifications
FrostedSandblasted
Length (Metric)76.2 mm
Thickness (Metric)0.97 to 1.07 mm
TypeMicroscope Slide
Width (Metric)25.4 mm
Length (English)3 in.
Width (English)1 in.
Unit SizePack