Self-Adhesive Labels
Self-Adhesive Labels
Actual product may vary
Fisher Catalogue
Self-Adhesive Labels
Use these white labels on a variety of surfaces. Fisherbrand™ Self-Adhesive Labels are removable and pressure sensitive. Ideal for a wide range of labeling applications.
Have Questions?
Catalog NumberColorQuantityLength (English)Width (English)
FIS11-870White, Blue1200/Pk.1 in.1 in.
Catalog number FIS11-870
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
White, Blue
Quantity:
1200/Pk.
Length (English):
1 in.
Width (English):
1 in.
Use these white labels on a variety of surfaces. Fisherbrand™ Self-Adhesive Labels are removable and pressure sensitive. Ideal for a wide range of labeling applications.
  • Self Adhesive Labels
  • Stick To Most Surfaces Without Moistening
  • Easily Marked With Pencil, Ballpoint Pen, or felt-tip Marker
Specifications
Adhesive TypeSelf-adhesive
ColorWhite, Blue
FormatLabel
MaterialPaper
Product TypeSelf Adhesive Label
For Use With (Application)General purpose
Quantity1200/Pk.
Dimensions (LxW)1 in. x 1 in.
Length (English)1 in.
Length (Metric)2.54 cm
Width (English)1 in.
Unit SizePK