Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Actual product may vary
Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English) Cuff
FIS11-394-29811 in.
FIS11-394-2995 in.
Catalog number FIS11-394-298
Price (MYR)
572.00
PK
Add to cart
Length (English) Cuff:
11 in.
Price (MYR)
572.00
PK
Add to cart
Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).
  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 232°C
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications
Certifications/ComplianceCE
Accelerator FreeYes
ApplicationsLaboratory, Industrial
Coating MaterialUncoated
ColorOrange
Latex FreeYes
MaterialTerrycloth
Product TypeAutoclave Gloves
SizeUniversal
StyleFull-hand Mitt
Temperature (English) Max.450°F
Temperature (Metric) Max232°C
For Use With (Application)Handling Hot Objects
Glove Cuff StyleGauntlet
SeriesH13201
Length (English)18.5 in.
Length (English) Cuff11 in.
Length (Metric)47 cm
Length (Metric) Cuff28 cm
Unit SizePK