Kimble™ KIMAX™ Solid Rod Stirrers
Kimble™ KIMAX™ Solid Rod Stirrers
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Solid Rod Stirrers
Designed from Federal Specification NNN-R-560, Type I requirements
Have Questions?
Catalog NumberDiameter (English)Diameter (Metric)Length (English)Length (Metric)
FIS11-381A0.15 in.0.38 cm4.9 in.125 mm
Catalog number FIS11-381A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English):
0.15 in.
Diameter (Metric):
0.38 cm
Length (English):
4.9 in.
Length (Metric):
125 mm
  • KG-33 borosilicate glass
  • Rounded ends
Specifications
Certifications/ComplianceUSP Type I, ASTM E438, Type I, Class A, NNN-R-560
Diameter (English)0.15 in.
Length (English)4.9 in.
Diameter (Metric)0.38 cm
Length (Metric)125 mm
MaterialBorosilicate Glass
Quantity800/Cs.
Unit SizePK