Soft Glass Rods
Soft Glass Rods
Actual product may vary
Fisher Catalogue
Soft Glass Rods
With rounded and fire-polished ends.
Have Questions?
Catalog NumberDiameter (English)Diameter (Metric)Length (English)Length (Metric)
FIS11-380D0.23 in.6 mm9.84 in.250 mm
Catalog number FIS11-380D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English):
0.23 in.
Diameter (Metric):
6 mm
Length (English):
9.84 in.
Length (Metric):
250 mm
  • Constructed of glass
  • A stirring rod is used to mix chemicals and liquids for laboratory purposes
Specifications
Diameter (English)0.23 in.
IncludesRounded and fire polished ends
Length (English)9.84 in.
Diameter (Metric)6 mm
Length (Metric)250 mm
MaterialGlass
QuantityCase of 1 pack (72 rods/pk.)
Unit SizePK