Soft Glass Stirring Rods
Soft Glass Stirring Rods
Actual product may vary
Fisher Catalogue
Soft Glass Stirring Rods
Available in different lengths ranging from 5 to 10 in.
Have Questions?
Catalog NumberDiameter (Metric)Length (Metric)
FIS11-380C4.5 mm203.2 mm
Catalog number FIS11-380C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
4.5 mm
Length (Metric):
203.2 mm
Available in different lengths ranging from 5 to 10 in.
  • With rounded and fire-polished ends
Specifications
Diameter (English)0.18 in.
Length (English)8 in.
Diameter (Metric)4.5 mm
Length (Metric)203.2 mm
MaterialGlass
QuantityCase of 72
TypeStirring Rod
Unit SizePK