Economical Solid Glass Beads for Distillation Columns
Economical Solid Glass Beads for Distillation Columns
Actual product may vary
Fisher Catalogue
Economical Solid Glass Beads for Distillation Columns
Used for mixing, boiling, or packing
Have Questions?
Catalog NumberDiameter (Metric)
FIS11-312A3 mm
Catalog number FIS11-312A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
3 mm
Used for mixing, boiling, or packing
  • Spherical Soda-lime glass
  • Chemical resistant
Specifications
TypeGlass Bead
MaterialGlass
Diameter (Metric)3 mm
For Use With (Equipment)Distillation Columns
Unit SizePK