Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe
Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe
Actual product may vary
Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe
Calibrated To Contain within Class A tolerances
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-248A25 mL
Catalog number FIS10-248A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
25 mL
  • Meet specifications in Class A Tolerances chart
  • Colorless graduation is clearly legible against contrasting red background
  • With standard taper pennyhead stoppers
Specifications
CalibrationCalibrated To Contain
Closure MaterialGlass
Capacity (Metric)25 mL
MaterialBorosilicate Glass
Height (English)4.64 in.
Height (Metric)11.8 cm
Tolerance±0.03 mL
GraduatedYes
Closure Size9
Unit SizeCS