Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Actual product may vary
Fisher Catalogue
Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Calibrated To Contain
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-212J1 mL
Catalog number FIS10-212J
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1 mL
Calibrated To Contain
  • Permanently marked with individual serial number
  • Individually calibrated and certified against equipment whose calibration is traceable to NIST™
  • Cylindrical body for better mixing and draining
  • Easier to withdraw samples using pipets
  • Wide base provides stability
  • Marking spot
  • Ground glass stopper
  • 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
stopper
Specifications
CalibrationCalibrated To Contain
Closure MaterialGlass
Capacity (Metric)1 mL
MaterialBorosilicate Glass
Height (English)3.7 in.
Height (Metric)9.4 cm
Tolerance±0.01 mL
GraduatedYes
Closure Size8
Unit SizeEA