PYREXPLUS™ Coated Heavy Wall Filtering Flasks with Sidearm Tubulation
Actual product may vary
PYREXPLUS™ Coated Heavy Wall Filtering Flasks with Sidearm Tubulation
Pyrex™ borosilicate glass with PVC coating reduces chance of breakage
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-180L1 L
Catalog number FIS10-180L
Price (MYR)
5,756.00
Case
Capacity (Metric):
1 L
Price (MYR)
5,756.00
Case
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. For convenience, the flasks are graduated to show approximate capacity. Tubulation O.D. is approximately 10 mm on sizes up to 1000 mL. Tubulation O.D. is approximately 12.5 mm on 2000 and 4000 mL sizes. The tubulation is not coated to allow easy connection of standard size tubing.
Do not place over direct heat or flame. Do not heat above 121°C moist heat or 110°C dry heat.
Specifications
DescriptionStyle: Filtering; Top style: Heavy duty rim
Diameter (English) Outer9.37 in.
Diameter (Metric) Outer Tubing1 cm
Capacity (Metric)1 L
MaterialBorosilicate Glass
Graduation Range300 to 1000 mL
Graduations50 mL
Height (English)14.01 in.
Height (Metric)35.6 cm
Neck StyleNarrow Mouth
No. per Case6/Cs.
No. per Pack6/Pk.
Surface CoatingPyrexplus
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer238 mm
Closure Size8
Unit SizeCase