PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
Actual product may vary
Fisherbrand
PYREX™ Narrow Mouth Erlenmeyer Flask with Standard Taper Stopper
Narrow mouths
Have Questions?
Catalog NumberCapacity (Metric)Material
FIS10-098E250 mLBorosilicate Glass
Catalog number FIS10-098E
Price (MYR)
2,237.00
PK
Add to cart
Capacity (Metric):
250 mL
Material:
Borosilicate Glass
Price (MYR)
2,237.00
PK
Add to cart
For convenience, these flasks are graduated to show approximate capacity. The 25 mL, 50 mL, and 125 mL flasks are satisfactory for use as a weighing bottle.
For stopper only, see Cat. No. 7650.
Specifications
Diameter (English) Outer3.18 in.
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Graduation Range50 to 200 mL
Graduations25 mL
Height (English)6.73 in.
Height (Metric)17 cm
Neck StyleNarrow Mouth
No. per Case12/Cs.
No. per Pack6/Pk.
Writing SurfaceYes
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Closure TypeBarrel Head
Diameter (Metric) Outer81 mm
Closure Size27
Unit SizePK