PYREX™ Narrow Mouth Heavy-Duty Glass Erlenmeyer Flask
PYREX™ Narrow Mouth Heavy-Duty Glass Erlenmeyer Flask
Actual product may vary
Fisherbrand
PYREX™ Narrow Mouth Heavy-Duty Glass Erlenmeyer Flask
Constructed of PYREX™ for durability. Erlenmeyer style, rugged tooled top finish and narrow mouth.
Have Questions?
Catalog NumberCapacity (Metric)Includes
FIS10-040C50 mLNo Stopper
Catalog number FIS10-040C
Price (MYR)
733.00
PK
Add to cart
Capacity (Metric):
50 mL
Includes:
No Stopper
Price (MYR)
733.00
PK
Add to cart
Constructed of PYREX™ for durability. Erlenmeyer style, rugged tooled top finish and narrow mouth.
Use these flasks for a variety of applications. PYREX™ Narrow Neck Heavy-Duty Erlenmeyer Flasks are designed for longer life and increased user safety.
Specifications
DescriptionTop style: Reinforced tooled
Diameter (English) Outer2 in.
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Graduation Range20 to 50 mL
Graduations10 mL
Height (English)3.07 in.
Height (Metric)7.8 cm
IncludesNo Stopper
Neck StyleNarrow Mouth
No. per Case48/Cs.
No. per Pack12/Pk.
Writing SurfaceLarge Enamel Marking
DisposableReusable
Mouth StyleNarrow
SterilityNon-sterile
Diameter (Metric) Outer51 mm
Closure Size1
Unit SizePK