Quantitative Grade Filter Paper Circles
Quantitative Grade Filter Paper Circles
Actual product may vary
Quantitative Grade Filter Paper Circles
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Particle Retention
FIS09-790G18.5 cm20 to 25 μm
FIS09-790-2G18.5 cm5 to 10 μm
FIS09-790-2F15 cm5 to 10 μm
FIS09-790-2E12.5 cm5 to 10 μm
FIS09-790-4E12.5 cm1 to 5 μm
FIS09-790-4F15 cm1 to 5 μm
FIS09-790E12.5 cm20 to 25 μm
FIS09-790F15 cm20 to 25 μm
Catalog number FIS09-790G
Price (MYR)
-
Diameter (Metric):
18.5 cm
Particle Retention:
20 to 25 μm
Manufactured to reduce silicon dioxide, calcium oxide and ferric oxide levels
  • Made from high-quality alpha cotton cellulose, washed in hydrochloric acid and neutralized in an ultrapure water rinse
  • Leave no more than 0.01% ash when burned
  • Limited wet strength, suitable for gravity or low-suction filtration; should be used with care to avoid rupturing
Specifications
FormatDisc
MaterialAlpha Cotton Cellulose
Particle Retention20 to 25 μm
Flow RateFast
For Use With (Application)Suitable for coarse and gelatinous precipitates requiring thorough washing on paper. Used in metallurgical labs for filtering iron and aluminum hydroxides, metallic aerosols and for determining silica in steel. Also used in environmental monitoring. Limited wet strength.
GradeQ8
PorosityCoarse
Quantity100/Pk.
Diameter (Metric)18.5 cm
Unit SizePK