Dissecting Cushing Delicate Thumb Forceps
Dissecting Cushing Delicate Thumb Forceps
Actual product may vary
Dissecting Cushing Delicate Thumb Forceps
Dissecting Cushing Delicate Thumb Forceps
Dissecting Cushing Delicate Thumb Forceps
Fisher Catalogue
Dissecting Cushing Delicate Thumb Forceps
Fisherbrand Dissecting Cushing Delicate Thumb Forceps are made from ASTM™ stainless steel construction. These forceps are light/medium weight and are primarily used to handle delicate tissue.
Have Questions?
Catalog number FIS08-953C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Fisherbrand™ Dissecting Cushing Delicate Thumb Forceps are primarily used to handle delicate tissue
  • Our forceps are designed to exceed performance expectations -This is the Fisherbrand Advantage
  • Multipurpose - Useful in educational, research or healthcare setting
  • Delicate, straight tips with serrated jaws
  • Delicate- very easy to press together to avoid nerve pain with controlled handling
  • Patterned grip for easy and comfortable handling
  • Durable and resists rusting
  • Hand polished to mirror finish
  • Made from stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully Autoclavable
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Product TypeDissecting Forcep
AutoclavableAutoclavable
Certifications/ComplianceASTM, FDA
MaterialStainless Steel
Latex FreeYes
Tip StyleStraight, serrated
Length (English)7 in.
Length (Metric)17.8 cm
Unit SizeEA