Stainless Steel Desiccator with Stainless Steel Shelves
Actual product may vary
Fisherbrand
Stainless Steel Desiccator with Stainless Steel Shelves
One piece neoprene gasket provides excellent sealing of the chamber.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS08-645-11Large
FIS08-645-6Standard
Catalog number FIS08-645-11
Price (MYR)
23,056.00
Each
Size:
Large
Price (MYR)
23,056.00
Each
One piece neoprene gasket provides excellent sealing of the chamber.
Key Features
  • Polished stainless steel with 0.187 in. thick (4.8mm) glass
  • Airtight door with neoprene gasket, full-height hinges and double-action bar locks
  • Fixed wire shelf brackets every 0.5 in. (13mm)
Standard Size
  • Glass on door and sides
  • Tray holds 454g of desiccant
  • Weighs 23 lb. (10kg)
Large Size
  • Glass on door and back, stainless-steel sides
  • Shelf dimensions: 13.375 x 18 in. (34 x 46cm)
  • Weighs 57 lb. (26kg)
Specifications
MaterialStainless Steel
Product TypeDesiccator Cabinet
IncludesAirtight door with neoprene gasket, full-height hinges, double-action bar locks, two stainless steel shelves and desiccant tray
No. of Shelves2
SizeLarge
Dimensions (L x W x H)22.5 x 16.5 x 23 in. (56.5 x 41.9 x 58 cm)
Length (English)23 in.
Length (Metric)584.2 mm
Width (English)16.5 in.
Width (Metric)41.9 cm
Height (English)22.25 in.
Height (Metric)565.15 mm
Weight (English)57 lb.
Weight (Metric)26 kg
Unit SizeEach