Check Order Status
Desiccator with Porcelain Plate
Actual product may vary
Fisherbrand
Desiccator with Porcelain Plate
Desiccant-holding chamber has straight sides.
Have Questions?
Catalog Number
Height (Metric)
Size
FIS08-615B
324 mm
250 mm (Inner Dia.)
Catalog number
FIS08-615B
Price (MYR)
2,928.00
EA
Add to cart
Height (Metric):
324 mm
Size:
250 mm (Inner Dia.)
Price (MYR)
2,928.00
EA
Add to cart
Desiccant-holding chamber has straight sides
Heavy glass construction
Sample plate has 5mm center hole, glaze on one side, and no feet
Cover has ground-on knob
Specifications
Material
Glass
Product Type
Nonvacuum Desiccator
Includes
Cover and porcelain sample plate with 0.5cm center hole, glaze on one side and no feet
Size
250 mm (Inner Dia.)
Height (Metric)
324 mm
Diameter (Metric) Inner
250 mm