Check Order Status
Fused Quartz Crucible Covers
Actual product may vary
Fisherbrand
Fused Quartz Crucible Covers
For 08-072 Series Crucibles
Have Questions?
Catalog Number
For Use With (Application)
Length (Metric)
FIS08-074D
For 30mL crucible
1cm
Catalog number
FIS08-074D
Price (MYR)
-
For Use With (Application):
For 30mL crucible
Length (Metric):
1cm
Transparent fused quartz
Specifications
Color
Clear, Transparent
Material
Quartz
For Use With (Application)
For 30mL crucible
Length (Metric)
1cm
Product Type
Crucible Lid