Castaloy™ Double-Buret Clamp
Castaloy™ Double-Buret Clamp
Actual product may vary
Fisherbrand
Castaloy™ Double-Buret Clamp
Holds two burets from micro to 100mL size
Have Questions?
Catalog number FIS05-779Q
Price (MYR)
-
Holds two burets from micro to 100mL size
Simply compress scissor-like mechanism, insert buret, and release.
  • Numbers and graduations on burets are easy to read
  • For height adjustments, just recompress mechanism, slide buret up or down, release Clamp unit attaches to standard support rod with convenient hook clamp
  • Wingnut for tightening on front for easy access
Specifications
Length (Metric)25.4 cm
For Use With (Equipment)Burette
MaterialCastaloy
For Use With (Application)Holding two burets from micro to 100mL size
Length (Metric) Arm25.4 cm
Length (English)10 in.
Length (English) Arm10 in.
TypeDouble Buret Clamp
Unit SizeEA