Check Order Status
Castaloy™ Clamp Regular Holder
Actual product may vary
Fisherbrand
Castaloy™ Clamp Regular Holder
Attaches clamp at right angles
Have Questions?
Catalog Number
Size (Metric) Grip
FIS05-754Q
19 mm
Catalog number
FIS05-754Q
Price (MYR)
-
Size (Metric) Grip:
19 mm
Attaches clamp at right angles
With 45° angle thumbscrews
Grips rods up to 0.75 in. (19mm) diameter
Specifications
Quantity
1 Ea.
Type
Regular Clamp Holder
Material
Die Cast Aluminum
Includes
45° Angle thumbscrews
For Use With (Equipment)
Rods up to 19 mm (0.75 in.) dia.
For Use With (Application)
Attaching clamp at right angle
Size (Metric) Grip
19 mm
Size (English) Grip
0.75 in.