Tube Brush
Tube Brush
Actual product may vary
Fisherbrand
Tube Brush
Tapered brush for 15mL centrifuge tubes.
Have Questions?
Catalog NumberColor
FIS03-608Tan
Catalog number FIS03-608
Price (MYR)
89.00
PK
Add to cart
Color:
Tan
Price (MYR)
89.00
PK
Add to cart
For 15mL centrifuge tubes.
  • Tapered bristle portion, 0.5 in. dia. (1.3cm) at tip
  • Galvanized wire handle
Specifications
Product TypeCentrifuge Tube Brush
Bristle MaterialNatural Hair
Handle MaterialGalvanized Wire
Tip TypeTapered
ColorTan
Diameter (English) Bristle0.81 in.
Diameter (Metric) Bristle2 cm
For Use With (Equipment)15mL centrifuge tubes
Length (English)9 in.
Length (English) Bristle3.5 in.
Length (Metric)23 cm
Length (Metric) Bristle8.9 cm
SizeSmall
Unit SizePK