Microscope Slide Boxes with Numbered Slots
Microscope Slide Boxes with Numbered Slots
Actual product may vary
Fisherbrand
Microscope Slide Boxes with Numbered Slots
Store and transport 25 or 100 slides safely with durable hinged cases. Fisherbrand™ Microscope Slide Boxes with Numbered Slots keeps standard microscope slides in secure slots that correspond to the numbered identification card. Stackable boxes close securely with latch lock.
Have Questions?
Catalog NumberColorSlide Capacity
FIS03-446Taupe100
Catalog number FIS03-446
Price (MYR)
48.00
EA
Add to cart
Color:
Taupe
Slide Capacity:
100
Price (MYR)
48.00
EA
Add to cart
Store and transport 25 or 100 slides safely with durable hinged cases. Fisherbrand™ Microscope Slide Boxes with Numbered Slots keeps standard microscope slides in secure slots that correspond to the numbered identification card. Stackable boxes clos
  • Constructed of durable polypropylene or paperboard, with no lining
  • Hinged lids are held securely closed by latch lock
  • Stacking feet provide maximize stability
  • Numbered slide identification card included for easy identification
Specifications
Slide Capacity100
Closure TypeHinged lid with latch lock
ColorTaupe
Device Holds100 Slides
Dimensions (L x W x H)210 x 169 x 37 mm
For Use With (Equipment)Standard Microscope Slides (25 x 75 mm)
Height (English)1.45 in.
Height (Metric)37 mm
IncludesNumbered index card
Length (English)8.26 in.
Length (English) Slide3 in.
Length (Metric)210 mm
Length (Metric) Slide75 mm
MaterialPolypropylene
StackableYes
Width (English)6.65 in.
Width (English) Slide1 in.
Width (Metric)169 mm
Width (Metric) Slide25 mm
Unit SizeEA