Borosilicate Glass Scintillation Vials, with White Polypropylene Caps
Borosilicate Glass Scintillation Vials, with White Polypropylene Caps
Actual product may vary
Fisherbrand
Borosilicate Glass Scintillation Vials, with White Polypropylene Caps
Use these general purpose vials with all common counters. Fisherbrand™ 20mL Borosilicate Glass Scintillation Vials with white polypropylene caps feature potassium-free glass composition which provides consistently low activity counts and excellent light transmission.
Have Questions?
Catalog NumberClosure Type
FIS03-337-4Attached Screw Cap
Catalog number FIS03-337-4
Price (MYR)
2,065.00
CS
Add to cart
Closure Type:
Attached Screw Cap
Price (MYR)
2,065.00
CS
Add to cart
Use these general purpose vials with all common counters. Fisherbrand™ 20mL Borosilicate Glass Scintillation Vials with white polypropylene caps feature potassium-free glass composition which provides consistently low activity counts and excellent light transmission.
  • Caps fit GPI 22-400 thread finish and are suitable for marking
Specifications
Material (Liner)Pulp-backed Foil
Diameter (English) Outer1.12 in.
ColorWhite
Diameter (Metric) Outer28 mm
MaterialBorosilicate Glass
Volume (Metric)20 mL
Height (English)2.37 in.
Packaging TypeShrink-wrapped Tray
Volume (English)0.67 oz.
Volume (Metric) Total20 mL
Closure MaterialPolypropylene
Closure TypeAttached Screw Cap
Height (Metric)61 mm
Quantity100 Cases (Vial), 100 Pk. (Cap)
TypeScintillation Vial
Closure Size22-400
Marking SpotYes
Unit SizeCS