Beaker Tongs with Fiberglas-Covered Jaws
Beaker Tongs with Fiberglas-Covered Jaws
Actual product may vary
Fisherbrand
Beaker Tongs with Fiberglas-Covered Jaws
Handle beakers from 100 to 1500mL
Have Questions?
Catalog NumberLength (Metric)
FIS02-62033 cm
Catalog number FIS02-620
Price (MYR)
305.00
EA
Add to cart
Length (Metric):
33 cm
Price (MYR)
305.00
EA
Add to cart
Handle beakers from 100 to 1500mL
  • Fisherbrand™ Beaker Tongs with Fiberglas™ - Covered Jaws. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Designed for safe handling of beakers and vessels from 100 mL to 1500 mL
  • Precision riveted joint with Fiberglas™-covered cupped jaws
  • Fiberglass sleeves for use in high-temperature applications
  • Made from ASTM™ standard stainless steel
Specifications
For Use With (Application)Safely handling 100 to 1500mL beakers
IncludesFiberglass covered jaws
Length (English)13 in.
Length (Metric)33 cm
MaterialStainless Steel
TypeBeaker Tongs
Unit SizeEA