PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Actual product may vary
Fisher Catalogue
PYREX™ Watch Glass/Beaker Cover with Fire-Polished Edges
Molded for table stacking
Have Questions?
Catalog NumberDiameter (Metric)For Use With (Equipment)
FIS02-612B7.5 cmFor 250 mL beaker
Catalog number FIS02-612B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
7.5 cm
For Use With (Equipment):
For 250 mL beaker
Molded for table stacking
These watch glasses possess high chemical durability, thus preventing contamination of solutions. They are molded to the same radius of curvature for convenient stacking. The heavy wall and ­uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
Specifications
DisposableReusable
MaterialBorosilicate Glass
ShapeRound
TypeWatch Glass
For Use With (Application)Used as Beaker Covers
For Use With (Equipment)For 250 mL beaker
Interior FinishPolished
No. per Case144/Cs.
No. per Pack12/Pk.
Diameter (English)2.95 in.
Diameter (Metric)7.5 cm
Unit SizePK