Urea, Certified AR for Analysis, Fisher Chemical™
Urea, Certified AR for Analysis, Fisher Chemical™
Urea, Certified AR for Analysis, Fisher Chemical™
Urea, Certified AR for Analysis, Fisher Chemical™
Urea, Certified AR for Analysis, Fisher Chemical™
CH4N2O, CAS Number-57-13-6, carbamimidic acid, alphadrate, carbonyldiamide, isourea, urevert, ureaphil, pseudourea, ureophil, carbonyldiamine, carbamide, 5kg, CHEBI:48376, White, 60.06g/mol, XSQUKJJJFZCRTK-UHFFFAOYSA-N, 131 deg.C, 8022, 60.056, Plastic keg, 7.5 to 9.5, Solid, 1176, C(-O)(N)N
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
3 kg
Packaging:
View all
Plastic keg
Catalog number FGCU050065
also known as FGCU/0500/65
Price (MYR)
861.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic keg
Request bulk or custom format
Price (MYR)
861.00
EA
Add to cart
Specifications
CAS57-13-6
Melting Point131°C
ColorWhite
pH7.5 to 9.5
PackagingPlastic keg
View more
CH4N2O, CAS Number-57-13-6, carbamimidic acid, alphadrate, carbonyldiamide, isourea, urevert, ureaphil, pseudourea, ureophil, carbonyldiamine, carbamide, 5kg, CHEBI:48376, White, 60.06g/mol, XSQUKJJJFZCRTK-UHFFFAOYSA-N, 131 deg.C, 8022, 60.056, Plastic keg, 7.5 to 9.5, Solid, 1176, C(-O)(N)N