Check Order Status
Urea, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Urea, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCU045053
also known as FGCU/0450/53
500 g
Catalog number
FGCU045053
also known as FGCU/0450/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS
57-13-6
Melting Point
131°C
Color
White
pH
7.5 to 9.5
Packaging
Plastic powder jar
View more