Check Order Status
Thymolphthalein, Pure, Solution 0.2%, Indicator Grade, Fisher Chemical
Element Search
Structure Search
Thymolphthalein, Pure, Solution 0.2%, Indicator Grade, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCT1480L08
also known as FGCT/1480L/08
500 mL
Catalog number
FGCT1480L08
also known as FGCT/1480L/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
MDL Number
5909
Packaging
Amber glass bottle
Color
Colorless
pH
9.3 to 10.5
Quantity
500 mL
View more