Check Order Status
Thymol Blue, Pure, Indicator Grade, Fisher Chemical
Element Search
Structure Search
Thymol Blue, Pure, Indicator Grade, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCT147044
also known as FGCT/1470/44
5 g
Catalog number
FGCT147044
also known as FGCT/1470/44
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Melting Point
221°C
CAS
76-61-9
Molecular Formula
C27H30O5S
MDL Number
5869
InChI Key
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
View more