Check Order Status
Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCT1000PB17
also known as FGCT/1000/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCT1000PB17
also known as FGCT/1000/PB17
Price (MYR)
-
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
CAS
68-11-1
Melting Point
-16°C
Color
Colorless
pH
1.5
Boiling Point
220°C
View more