Check Order Status
Tetrahydrofuran, Gel Permeation Chromatography (GPC), Stabilised with 0.025% BHT, Fisher Chemical
Element Search
Structure Search
Tetrahydrofuran, Gel Permeation Chromatography (GPC), Stabilised with 0.025% BHT, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCT0709PB17
also known as FGCT/0709/PB17
2.5 L
Plastic-coated Glass Bottle
Catalog number
FGCT0709PB17
also known as FGCT/0709/PB17
Price (MYR)
482.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
482.00
BOT
Add to cart
Specifications
pH
7 to 8
Melting Point
-108.4°C
CAS
109-99-9
Color
Colorless
Boiling Point
66°C
View more