▸
Lab Desiccants and Drying Agents
▸
Silica gel (2.5-6.0mm) self indicating, dessicant agent, Fisher Chemical™
Element Search
Structure Search
Silica gel (2.5-6.0mm) self indicating, dessicant agent, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic Keg
Catalog number
FGCS076265
also known as FGCS/0762/65
Price (MYR)
1,321.00
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic Keg
Request bulk or custom format
Price (MYR)
1,321.00
Each
Add to cart
Specifications
Color
Orange
Melting Point
1710°C
Physical Form
Crystalline Solid
pH
2.0 to 3.5
CAS
7631-86-9
View more