Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Actual product may vary
Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FGCS076253
also known as FGCS/0762/53
500 gPlastic Powder Jar
Catalog number FGCS076253
also known as FGCS/0762/53
Price (MYR)
350.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic Powder Jar
Price (MYR)
350.00
EA
Add to cart
500g, Plastic powder jar
Specifications
ColorWhite
Physical FormSolid
pH2.0 to 3.5
CAS7631-86-9
Quantity500 g
Molecular FormulaO2Si
MDL NumberMFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
PackagingPlastic Powder Jar
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
IUPAC Namedioxosilane
Molecular Weight (g/mol)60.08
PubChem CID24261
ChEBICHEBI:30563
Formula Weight60.08g/mol
Unit SizeEA