▸
Lab Desiccants and Drying Agents
▸
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Element Search
Structure Search
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCS076153
also known as FGCS/0761/53
500 g
Plastic Powder Jar
Catalog number
FGCS076153
also known as FGCS/0761/53
Price (MYR)
378.00
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic Powder Jar
Request bulk or custom format
Price (MYR)
378.00
Each
Add to cart
Specifications
Color
Brown-Orange
Physical Form
Solid
pH
1.5 to 5
CAS
7631-86-9
Quantity
500 g
View more
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent