Check Order Status
Rhodizonic Acid Sodium Salt, Pure, Fisher Chemical
Element Search
Structure Search
Rhodizonic Acid Sodium Salt, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCR030044
also known as FGCR/0300/44
5 g
Glass Jar
Catalog number
FGCR030044
also known as FGCR/0300/44
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass Jar
Request bulk or custom format
Specifications
Melting Point
300°C
CAS
523-21-7
Color
Green
Physical Form
Powder/Solid
Quantity
5 g
View more