Check Order Status
Molecular Sieve
Actual product may vary
Molecular Sieve
Molecular Sieve Type 3A | CAS: 308080-99-1
Have Questions?
Catalog Number
Quantity
FGCMS101050
also known as FGCMS/1010/50
250 g
Catalog number
FGCMS101050
also known as FGCMS/1010/50
Price (MYR)
341.00
EA
Add to cart
Quantity:
250 g
Price (MYR)
341.00
EA
Add to cart
Molecular Sieve Type 3A
Molecular Sieve
Type 3A
250 g
Nominal pore size 3
Angstroms Bead size 1.6-2.5 mm
CAS No. 308080-99-1
Specifications
Chemical Name or Material
Molecular Sieve
Packaging
Plastic
Quantity
250 g
CAS
308080-99-1