Check Order Status
Methyl Red, Pure, Water Soluble, Indicator, Fisher Chemical™
Element Search
Structure Search
Methyl Red, Pure, Water Soluble, Indicator, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM517048
also known as FGCM/5170/48
100 g
Catalog number
FGCM517048
also known as FGCM/5170/48
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Melting Point
178°C
CAS
493-52-7
Molecular Formula
C15H15N3O2
MDL Number
2425
InChI Key
CEQFOVLGLXCDCX-UHFFFAOYSA-N
View more