Check Order Status
Methyl Red, Solution 0.01%, Pure, Spirit Soluble, C.I 13020, Indicator Grade, Fisher Chemical
Element Search
Structure Search
Methyl Red, Solution 0.01%, Pure, Spirit Soluble, C.I 13020, Indicator Grade, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCM5150L08
also known as FGCM/5150L/08
500 mL
Catalog number
FGCM5150L08
also known as FGCM/5150L/08
Price (MYR)
495.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
495.00
EA
Add to cart
Specifications
MDL Number
2425
Packaging
Amber glass bottle
Quantity
500 mL
Physical Form
Liquid