Check Order Status
N-Methylimidazole, 99% (GLC), for DNA Synthesis, Fisher Chemical™
Element Search
Structure Search
N-Methylimidazole, 99% (GLC), for DNA Synthesis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCM4930PB17
also known as FGCM/4930/PB17
2.5 L
Catalog number
FGCM4930PB17
also known as FGCM/4930/PB17
Price (MYR)
4,216.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
4,216.00
EA
Add to cart
Specifications
CAS
616-47-7
Melting Point
-2°C
Color
Yellow
Boiling Point
198°C
Packaging
Plastic Coated Glass bottle
View more