Check Order Status
Karl Fischer Aqualine™ Water Standard 0.2mg, for Karl Fischer Titration, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Water Standard 0.2mg, for Karl Fischer Titration, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCK274099
also known as FGCK/2740/99
40 mL
Catalog number
FGCK274099
also known as FGCK/2740/99
Price (MYR)
450.00
EA
Add to cart
Quantity:
40 mL
Request bulk or custom format
Price (MYR)
450.00
EA
Add to cart
Specifications
Color
Colorless
Quantity
40 mL
Boiling Point
138.5°C
Packaging
Ampoule
Melting Point
-34°C
View more