Check Order Status
Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical
10mg/mL H
2
O standard
Have Questions?
Catalog Number
Quantity
FGCK272099
also known as FGCK/2720/99
10 x 8 mL
Catalog number
FGCK272099
also known as FGCK/2720/99
Price (MYR)
699.00
BOT
Add to cart
Quantity:
10 x 8 mL
Request bulk or custom format
Price (MYR)
699.00
BOT
Add to cart
Specifications
Chemical Name or Material
Aqualine™ Standard 10.1
Quantity
10 x 8 mL
Cleanup Symbol
Solvent
Name Note
Karl Fischer Reagent
CAS
71-36-3
View more
Standard contains 10mg H
2
O/mL
Used for titer determination of volumetric Karl Fischer reagents
Supplied as 10 x 8mL and packaged in glass ampules to protect from moisture
The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere