Check Order Status
Hyamine Solution 5% (w/v), SLR, Extra Pure, Fisher Chemical
Element Search
Structure Search
Hyamine Solution 5% (w/v), SLR, Extra Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ901615
also known as FGCJ/9016/15
1 L
Amber glass bottle
Catalog number
FGCJ901615
also known as FGCJ/9016/15
Price (MYR)
-
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS
121-54-0
Packaging
Amber glass bottle
Molecular Formula
C27H42ClNO2
MDL Number
11742
Quantity
1 L
View more