Check Order Status
Hydrochloric Acid Solution 5M (5N), Standard Solution ready to use for volumetric analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 5M (5N), Standard Solution ready to use for volumetric analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FGCJ431017
also known as FGCJ/4310/17
Price (MYR)
361.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
361.00
EA
Add to cart
Specifications
pH
lt; 1
Color
Colorless to Yellow
CAS
7647-01-0
Packaging
HDPE plastic bottle
Quantity
2.5 L
View more
Hydrochloric Acid Solution 5M (5N), Standard Solution ready to use for volumetric analysis