Check Order Status
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical
Element Search
Structure Search
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ3720C05
also known as FGCJ/3720C/05
1 Ampule
Ampoule
Catalog number
FGCJ3720C05
also known as FGCJ/3720C/05
Price (MYR)
272.00
EA
Add to cart
Quantity:
1 Ampule
Packaging:
Ampoule
Request bulk or custom format
Price (MYR)
272.00
EA
Add to cart
Specifications
CAS
139-33-3
Color
Colorless
Packaging
Ampoule
Molecular Formula
C10H14N2Na2O8
MDL Number
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
View more
For volumetric analysis