Check Order Status
Chromic Acid, Fisher Chemical
Element Search
Structure Search
Chromic Acid, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ3500PC15
also known as FGCJ/3500/PC15
1 L
Plastic Coated Glass bottle
Catalog number
FGCJ3500PC15
also known as FGCJ/3500/PC15
Price (MYR)
-
Quantity:
1 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
pH
0.3
Color
Colorless to Yellow
Melting Point
10°C
Boiling Point
290°C
Packaging
Plastic Coated Glass bottle
View more