Check Order Status
Acetic Acid, 2M (2N ≈12%), Pure, Ready To Use
Element Search
Structure Search
Acetic Acid, 2M (2N ≈12%), Pure, Ready To Use
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ0549PB17
also known as FGCJ/0549/PB17
2.5 L
HDPE plastic bottle
Catalog number
FGCJ0549PB17
also known as FGCJ/0549/PB17
Price (MYR)
873.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
873.00
EA
Add to cart
Specifications
CAS
64-19-7
Boiling Point
100°C
Color
Colorless
Molecular Formula
C2H4O2
MDL Number
MFCD00036152
View more