Check Order Status
Iron Oxide, Pure, Magnetic, Precipitated, Fisher Chemical
Element Search
Structure Search
Iron Oxide, Pure, Magnetic, Precipitated, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCI110053
also known as FGCI/1100/53
500 g
Glass Jar
Catalog number
FGCI110053
also known as FGCI/1100/53
Price (MYR)
444.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Jar
Request bulk or custom format
Price (MYR)
444.00
EA
Add to cart
Specifications
Melting Point
1588.9°C
CAS
1317-61-9
Color
Black
pH
4 to 8
Physical Form
Solid
View more