Check Order Status
N-Hexane, Certified AR for Analysis, Fisher Chemical™
Actual product may vary
N-Hexane, Certified AR for Analysis, Fisher Chemical™
C6H14, CAS Number-110-54-3, hexanes, hexyl hydride, hexan, hexanen, gettysolve-b, heksan, dipropyl, esani, n-hexane, skellysolve b, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 2179311, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97 | CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FGCH0421PB17
also known as FGCH/0421/PB17
2.5 L
Plastic-coated Glass Bottle
Catalog number
FGCH0421PB17
also known as FGCH/0421/PB17
Price (MYR)
1,073.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Price (MYR)
1,073.00
BOT
Add to cart
C6H14, CAS Number-110-54-3, hexanes, hexyl hydride, hexan, hexanen, gettysolve-b, heksan, dipropyl, esani, n-hexane, skellysolve b, 2.5L, 69 deg.C, CHEBI:29021, Colorless, 86.18g/mol, VLKZOEOYAKHREP-UHFFFAOYSA-N, -95 deg.C, 2179311, 86.178, Plastic Coated Glass bottle, Liquid, 8058, CCCCCC, 2.97
Specifications
Melting Point
-95°C
CAS
110-54-3
Color
Colorless
Boiling Point
69°C
Quantity
2.5 L
Packaging
Plastic-coated Glass Bottle
Physical Form
Liquid
Molecular Formula
C6H14
MDL Number
MFCD02179311
Vapor Pressure
160mbar at 20°C
Viscosity
0.31 mPaS at 20°C
Synonym
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
IUPAC Name
hexane
Molecular Weight (g/mol)
86.18
PubChem CID
8058
ChEBI
CHEBI:29021
Formula Weight
86.18g/mol