Check Order Status
Heptane, Extra Pure, Fraction, SLR, Fisher Chemical™
Element Search
Structure Search
Heptane, Extra Pure, Fraction, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCH010017
also known as FGCH/0100/17
2.5 L
Catalog number
FGCH010017
also known as FGCH/0100/17
Price (MYR)
375.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
375.00
EA
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
64742-49-0
Vapor Pressure
48mbar at 20°C
Viscosity
0.4 mPaS at 20°C
Vapor Density
3.5
View more
Heptane, Extra Pure, Fraction, SLR