Check Order Status
N-Heptafluorobutyryl-Imidazole, 98+%, Pure, Fisher Chemical
Element Search
Structure Search
N-Heptafluorobutyryl-Imidazole, 98+%, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCH009545
also known as FGCH/0095/45
5 g
Plastic container
Catalog number
FGCH009545
also known as FGCH/0095/45
Price (MYR)
-
Quantity:
5 g
Packaging:
Plastic container
Request bulk or custom format
Specifications
CAS
32477-35-3
Melting Point
10°C
Color
Colorless
Boiling Point
158°C
Packaging
Plastic container
View more